missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropylmethyleisothiocyanate, 97%, Thermo Scientific™

Product Code. 10457822
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10457822 1 g Amber Glass Bottle 1g
10697734 5 g Amber Glass Bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10457822 Supplier Thermo Scientific Maybridge Supplier No. S10467DA
 Restricted Item
Estimated Shipment: 18-09-2026
Add to basket
This item is not returnable. View return policy

Chemical Identifiers

CAS 6068-90-2
Molecular Formula C5H7NS
Molecular Weight (g/mol) 113.178
MDL Number MFCD00052446
InChI Key YRJONJBJAYHHQB-UHFFFAOYSA-N
Synonym cyclopropylmethyl isothiocyanate, isothiocyanatomethyl cyclopropane, cyclopropylmethylisothiocyanate, cyclopropane, isothiocyanatomethyl, cyclopropylmethanisothiocyanate
PubChem CID 2795403
IUPAC Name isothiocyanatomethylcyclopropane
SMILES C1CC1CN=C=S

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Linear Formula C3H5CH2NCS
Quantity 1 g
Formula Weight 113.18
Percent Purity 97%
Chemical Name or Material Cyclopropylmethyleisothiocyanate
compliance-icons compliance-icons
Product Identifier
  • Cyclopropylmethyl isothiocyanate
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H318-Causes serious eye damage.
  • H314-Causes severe skin burns and eye damage.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.