missing translation for 'onlineSavingsMsg'
Learn More

Cyclopropyltriphenylphosphonium bromide, 98%

Product Code. 11466127
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11466127

Brand: Thermo Scientific Alfa Aesar B25102.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14114-05-7 | C21H20BrP | 383.27 g/mol

It is used for electrolytic polarization of Zn, has potential antitumor activity, reactant for synthesis of cycloporpylidenemethylarylethynynes, keto halides for sequential azidation and intramolecular Schmidt reactions, a reactant for hydroalkynylation.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used for electrolytic polarization of Zn, has potential antitumor activity, reactant for synthesis of cycloporpylidenemethylarylethynynes, keto halides for sequential azidation and intramolecular Schmidt reactions, a reactant for hydroalkynylation.

Solubility
Soluble in water.

Notes
Hygroscopic. Store under inert gas. Store away from oxidizing agents, water/ moisture.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14114-05-7
Molecular Formula C21H20BrP
Molecular Weight (g/mol) 383.27
MDL Number MFCD00011872
InChI Key XMPWFKHMCNRJCL-UHFFFAOYSA-M
Synonym cyclopropyltriphenylphosphonium bromide, phosphonium, cyclopropyltriphenyl-, bromide, cyclopropyltriphenylphosphanium bromide, cyclopropyl triphenyl phosphanium bromide, acmc-209cls, cyclopropyltriphenylphosphoniumbromide, cyclopropyl triphenylphosphonium bromide, cyclopropyl-triphenylphosphonium bromide, cyclopropyltriphenyl-phosphonium bromide, triphenyl-cyclopropylphosphonium bromide
PubChem CID 2723931
IUPAC Name cyclopropyl(triphenyl)phosphanium;bromide
SMILES [Br-].C1CC1[P+](C1=CC=CC=C1)(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 183°C to 186°C
Quantity 100 g
Beilstein 6413483
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 383.27
Percent Purity 98%
Chemical Name or Material Cyclopropyltriphenylphosphonium bromide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.