missing translation for 'onlineSavingsMsg'
Learn More

Cyclosporin A, MedChemExpress

Product Code. 16471426
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16471426 200 mg 200mg
16438376 100 mg 100mg
16462176 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16471426 Supplier Medchem Express Supplier No. HYB0579
 Restricted Item
Estimated Shipment: 28-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress Cyclosporin A (Cyclosporine A) is an immunosuppressant which binds to the cyclophilin and inhibits phosphatase activity of calcineurin with an IC50 of 5 nM. Cyclosporin A also inhibits CD11a/CD18 adhesion.

Cyclosporin A (Cyclosporine A) is an immunosuppressant which binds to the cyclophilin and inhibits phosphatase activity of calcineurin with an IC50 of 5 nM. Cyclosporin A also inhibits CD11a/CD18 adhesion.

Specifications

Chemical Name or Material Cyclosporin A
Physical Form Solid
CAS 59865-13-3
Color White
Quantity 200 mg
Purity Grade Notes Research
Molecular Formula C62H111N11O12
For Use With (Application) COVID-19-immunoregulation
Synonym Cyclosporine A Ciclosporin A CsA
Solubility Information DMSO : 62.5 mg/mL (51.97 mM; Need ultrasonic) ∣Ethanol : 50 mg/mL (41.58 mM; Need ultrasonic)
SMILES C/C=C/C[C@H]([C@@H](O)[C@@]1(N(C)C([C@]([H])(C(C)C)N(C)C([C@H](CC(C)C)N(C)C([C@]([H])(CC(C)C)N(C)C([C@@H](C)NC([C@H](C)NC([C@@H](N(C([C@@]([H])(NC([C@@H](N(C(CN(C([C@@H](NC1=O)CC)=O)C)=O)C)CC(C)C)=O)C(C)C)=O)C)CC(C)C)=O)=O)=O)=O)=O)=O)[H])C
Molecular Weight (g/mol) 1202.61
Formula Weight 1202.61
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.