missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CYFIP1 Recombinant Rabbit Monoclonal Antibody (23GB1605), Invitrogen™

Rabbit Recombinant Monoclonal Antibody
207.00 € - 562.00 €
Specifications
| Antigen | CYFIP1 |
|---|---|
| Clone | 23GB1605 |
| Applications | Flow Cytometry, Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
30088898
![]() |
Invitrogen™
MA553852 |
100 μL |
562.00 €
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30089230
|
Invitrogen™
MA553853 |
20 μL |
207.00 €
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CYFIP1 Recombinant Monoclonal antibody specifically detects CYFIP1 in Human, Mouse, Rat samples. It is validated for Flow Cytometry, Immunocytochemistry, Western BlotSpecifications
| CYFIP1 | |
| Flow Cytometry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| Q7L576, Q7TMB8 | |
| 20430, 23191, 308666 | |
| IgG | |
| Affinity chromatography | |
| Antibody |
| 23GB1605 | |
| Recombinant Monoclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| CY; CYFIP; cyfip1; cyfip1 protein; cytoplasmic FMR1 interacting protein 1; cytoplasmic FMR1-interacting protein 1; Cytoplasmic FMR1-interacting protein 1 homolog; cytoplasmic FMRP interacting protein 1; E030028J09Rik; fb15a06; FLJ45151; FMR1-interacting protein 1; KIAA0068; l(7)1Rl; l71Rl; l7Rl1; LOW QUALITY PROTEIN: cytoplasmic FMR1-interacting protein 1; mKIAA0068; P140sra1; p140sra-1; pl-1; selective hybridizing clone; Shyc; Specifically Rac1-associated protein 1; Sra1; Sra-1; Unknown (protein for IMAGE:7152255); wu:fb15a06 | |
| A synthesized peptide derived from human CYFIP1 (50-100AA). | |
| Primary | |
| -20°C | |
| Cyfip1 |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title