missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyflumetofen Metabolite AB-1 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | Cyflumetofen Metabolite AB-1 |
| CAS | 211923-03-4 |
| Analyte or Component Names | Cyflumetofen Metabolite AB-1 |
| Shipping Condition | +20°C |
| Molecular Formula | C20 H18 F3 N O |
| Concentration | 100 μg/mL |
| Certifications/Compliance | ISO 17034 |
| Synonym | 4-(1,1-Dimethylethyl)-α-[hydroxy[2-(trifluoromethyl)phenyl]methylene]benzeneacetonitrile, Cyflumetofen Metabolite AB-1 |
| InChI Formula | InChI=1S/C20H18F3NO/c1-19(2,3)14-10-8-13(9-11-14)16(12-24)18(25)15-6-4-5-7-17(15)20(21,22)23/h4-11,25H,1-3H3 |
| SMILES | CC(C)(C)c1ccc(cc1)C(=C(O)c2ccccc2C(F)(F)F)C#N |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?