missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYP11B1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-49223
This item is not returnable.
View return policy
Description
CYP11B1 Polyclonal antibody specifically detects CYP11B1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| CYP11B1 | |
| Polyclonal | |
| Immunohistochemistry 1:50 - 1:200, Immunohistochemistry-Paraffin 1:50 - 1:200 | |
| CPN1, CYPXIB1, cytochrome P450 11B1, mitochondrial, cytochrome p450 XIB1, cytochrome P450, family 11, subfamily B, polypeptide 1, cytochrome P450, subfamily XIB (steroid 11-beta-hydroxylase), polypeptide 1, Cytochrome P-450c11, Cytochrome P450C11, EC 1.14.15, EC 1.14.15.4, FHICYP11B, FLJ36771, P450C11DKFZp686B05283, S11BH, Steroid 11-beta-hydroxylase, steroid 11-beta-monooxygenase | |
| This antibody was developed against a recombinant protein corresponding to amino acids: RFLPMVDAVARDFSQALKKKVLQNARGSLTLDVQPSIFHYTIEASNLALFGERLGLVGHSPSSASLNFLHALEVMFKSTVQLMFMPR | |
| 0.1 mL | |
| Lipid and Metabolism | |
| 1584 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2), 40% Glycerol | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction