missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYP17A1 Monoclonal Antibody (OTI9H4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA503395S
This item is not returnable.
View return policy
Description
CYP17A1 Monoclonal antibody specifically detects CYP17A1 in Canine, Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| CYP17A1 | |
| Monoclonal | |
| 1.2 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O08749, P09622, P49819, Q6P6R2 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 13382, 1738, 298942, 403978 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Western Blot | |
| OTI9H4 | |
| Unconjugated | |
| Dld | |
| 17a-hydroxylase; 17-alpha-hydroxyprogesterone aldolase; CPT7; Cyp17; Cyp17a1; CYPXVII; cytochrome P450 17A1; cytochrome P450 family 17 subfamily A member 1; cytochrome p450 XVIIA1; cytochrome P450, 17; cytochrome P450, 17a1; cytochrome P450, family 17, subfamily A, polypeptide 1; cytochrome P450, subfamily 17; Cytochrome P450, subfamily XVII; cytochrome P450, subfamily XVII (steroid 17-alpha-hydroxylase), adrenal hyperplasia; Cytochrome P450c17; cytochrome P450-C17; HGNC:2593; P45017alpha; p450c17; P450-C17; S17AH; steroid 17-alpha hydroxylase; steroid 17-alpha-hydroxylase/17,20 lyase; Steroid 17-alpha-monooxygenase | |
| Full length human recombinant protein of human DLD produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Canine, Human, Mouse, Rat | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction