missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CYP2D6 Polyclonal Antibody
GREENER_CHOICE

Product Code. 13454087
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
13454087 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 13454087 Supplier Invitrogen™ Supplier No. PA529797

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Recommended positive controls: A431, HeLa, HepG2, Molt-4, Raji. Predicted reactivity: Cat (83%), Sheep (87%), Rhesus Monkey (96%), Chimpanzee (97%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Cytochrome p450 2D6 (CYP2D6) is a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This protein localizes to the endoplasmic reticulum and is known to metabolize as many as 20% of commonly prescribed drugs. Its substrates include debrisoquine, an adrenergic-blocking drug; sparteine and propafenone, both anti-arrythmic drugs; and amitryptiline, an anti-depressant. The gene is highly polymorphic in the population; certain alleles result in the poor metabolizer phenotype, characterized by a decreased ability to metabolize the enzyme's substrates. The gene is located near two cytochrome P450 pseudogenes on chromosome 22q13.1. Alternatively spliced transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CYP2D6
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene CYP2D6
Gene Accession No. P10635
Gene Alias AI303445; Cholesterol 25-hydroxylase; CPD6; CYP2D; Cyp2d10; Cyp2d-10; Cyp2d13; Cyp2d18; Cyp2d-18; Cyp2d22; Cyp2d3; Cyp2d-3; Cyp2d4; Cyp2d-4; Cyp2d4v1; Cyp2d4v2; Cyp2d6; CYP2D7AP; CYP2D7BP; CYP2D7P2; CYP2D8P2; CYP2DL1; CYPIID10; CYPIID18; CYPIID3; CYPIID4; CYPIID6; Cytochrome P450 2D10; cytochrome P450 2d18; Cytochrome P450 2D-29; Cytochrome P450 2D3; cytochrome P450 2D-35; cytochrome P450 2D4; Cytochrome P450 2D6; cytochrome P450 family 2 subfamily D member 6; cytochrome P450, 2d10; cytochrome P450, family 2, subfamily d, polypeptide 10; cytochrome P450, family 2, subfamily d, polypeptide 13; cytochrome P450, family 2, subfamily d, polypeptide 22; cytochrome P450, family 2, subfamily d, polypeptide 3; cytochrome P450, family 2, subfamily d, polypeptide 4; cytochrome P450, family 2, subfamily d, polypeptide 6; cytochrome P450, family 2, subfamily D, polypeptide 7 pseudogene 2; cytochrome P450, family 2, subfamily D, polypeptide 8 pseudogene 2; cytochrome P450, subfamily 2D, polypeptide 6; cytochrome P450, subfamily II (debrisoquine, sparteine, etc., -metabolising), polypeptide 7 pseudogene 2; cytochrome P450, subfamily IID (debrisoquine, sparteine, etc., -metabolising), polypeptide 8 pseudogene 2; cytochrome P450, subfamily IID (debrisoquine, sparteine, etc., -metabolizing), polypeptide 6; cytochrome P450, subfamily IID (debrisoquine, sparteine, etc., -metabolizing)-like 1; cytochrome P450, subfamily IID, polypeptide 6; Cytochrome P450, subfamily IID3; Cytochrome P450, subfamily IID4; cytochrome P450-16-alpha; Cytochrome P450CB; Cytochrome P450-CMF3; cytochrome P450-DB1; Cytochrome P450-DB3; cytochrome P450-DB4; debrisoquine 4-hydroxylase; flavoprotein-linked monooxygenase; microsomal monooxygenase; nonfunctional cytochrome P450 2D6; P450 2D-29/2D-35; P450-2D; P450C2D; P450-CMF3; P450DB1; P450-DB1; P450-DB3; P450-DB4; testosterone 16-alpha hydroxylase; xenobiotic monooxygenase
Gene Symbols CYP2D6
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human Cytochrome P450 2D6. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1565
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.