missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CYP3A7 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-21388-100ul
This item is not returnable.
View return policy
Description
CYP3A7 Polyclonal antibody specifically detects CYP3A7 in Human samples. It is validated for Western Blot
Specifications
| CYP3A7 | |
| Polyclonal | |
| Western Blot 0.04-0.4 ug/ml | |
| CYPIIIA7, cytochrome P450, family 3, subfamily A, polypeptide 7, Cytochrome P450-HFLA, EC 1.14.14.1, flavoprotein-linked monooxygenase, microsomal monooxygenase, subfamily IIIA, polypeptide 7, xenobiotic monooxygenase | |
| This antibody was developed against Recombinant Protein corresponding to amino acids: LATHPDVQQKVQKEIDTVLPNKAPPTYD | |
| 100 μg | |
| Lipid and Metabolism | |
| 1551 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS, pH 7.2, 40% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction