missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CYP7A1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA597650
This item is not returnable.
View return policy
Description
CYP7A1 Polyclonal Antibody for Western Blot, IHC (P), ELISA
CYP7A1 is a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This endoplasmic reticulum membrane protein catalyzes the first reaction in the cholesterol catabolic pathway in the liver, which converts cholesterol to bile acids. This reaction is the rate limiting step and the major site of regulation of bile acid synthesis, which is the primary mechanism for the removal of cholesterol from the body.
Specifications
| CYP7A1 | |
| Polyclonal | |
| Unconjugated | |
| Cyp7a1 | |
| 24-hydroxycholesterol 7-alpha-hydroxylase; CHAP; cholesterol 7 alpha hydroxylase; cholesterol 7-alpha hydroxylase; cholesterol 7-alpha-hydroxylase; Cholesterol 7-alpha-monooxygenase; CP7A; CYP7; Cyp7a1; CYP7S1; CYPVII; cytochrome P450 (cholesterol hydroxylase 7 alpha); cytochrome P450 7A1; cytochrome P450 family 7 subfamily A member 1; cytochrome P450, 7a1; cytochrome P450, family 7, subfamily a, polypeptide 1; cytochrome P450, subfamily VIIA polypeptide 1 | |
| Rabbit | |
| Protein G, Antigen affinity chromatography | |
| RUO | |
| 1581 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot | |
| 4.5 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300, 0.02% sodium azide; pH 7.4 | |
| P22680 | |
| Cyp7a1 | |
| Recombinant Human Cholesterol 7-alpha-monooxygenase protein (303-423AA). | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction