missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyr61/CCN1 Antibody (3A7.1B8), Biotin, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-36490B
This item is not returnable.
View return policy
Description
Cyr61/CCN1 Monoclonal specifically detects Cyr61/CCN1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| Cyr61/CCN1 | |
| Monoclonal | |
| Biotin | |
| PBS with 0.05% Sodium Azide | |
| CCN family member 1, CCN1IGF-binding protein 10, Cysteine-rich angiogenic inducer 61, cysteine-rich, angiogenic inducer, 61, GIG1cysteine-rich heparin-binding protein 61, IBP-10, IGFBP-10, IGFBP10cysteine-rich, anigogenic inducer, 61, Insulin-like growth factor-binding protein 10, protein CYR61, Protein GIG1 | |
| Mouse | |
| Protein G purified | |
| RUO | |
| 3491 | |
| Store at 4C in the dark. | |
| IgG2b κ |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 3A7.1B8 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| O00622 | |
| CCN1 | |
| Partial recombinant protein from human Cyr61/CCN1 protein (between residues 100-300). [Swiss-Prot O00622] | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction