missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cyromazine 100 μg/mL in Acetonitrile, Dr. Ehrenstorfer
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
Specifications
Specifications
| Chemical Name or Material | Cyromazine |
| CAS | 66215-27-8 |
| Analyte or Component Names | Cyromazine |
| Shipping Condition | Room Temperature |
| Molecular Formula | C6 H10 N6 |
| Concentration | 100 μg/mL |
| Certifications/Compliance | ISO 17034 |
| InChI Formula | InChI=1S/C6H10N6/c7-4-10-5(8)12-6(11-4)9-3-1-2-3/h3H,1-2H2,(H5,7,8,9,10,11,12) |
| SMILES | Nc1nc(N)nc(NC2CC2)n1 |
| IUPAC Name | 4-N-cyclopropyl-1,3,5-triazine-2,4,6-triamine |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction