missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cystatin F Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-99007-50ul
This item is not returnable.
View return policy
Description
Cystatin F Polyclonal antibody specifically detects Cystatin F in Human samples. It is validated for Western Blot
Specifications
| Cystatin F | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| CMAP, cystatin F (leukocystatin), Cystatin-7, cystatin-F, Cystatin-like metastasis-associated protein, leukocystatin | |
| Produced in rabbits immunized with purified, recombinant Human Cystatin F (Accession#: NP_003641.3; Met1-His145) | |
| 50 μg | |
| Angiogenesis, Apoptosis, Cardiovascular Biology, Cell Biology | |
| 8530 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| 0.2 μm filtered solution in PBS | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction