missing translation for 'onlineSavingsMsg'
Learn More

Cysteamine hydrochloride, MedChemExpress

Product Code. 16490668
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
Unit Size:
5g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16490668 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16490668 Supplier Medchem Express Supplier No. HY77591
 Restricted Item

This item requires a price update prior to being ordered.

Request a Price for this item here.

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Cysteamine hydrochloride (2-Aminoethanethiol hydrochloride) is an orally active agent for the treatment of nephropathic cystinosis and an antioxidant.

Cysteamine hydrochloride (2-Aminoethanethiol hydrochloride) is an orally active agent for the treatment of nephropathic cystinosis and an antioxidant.

Specifications

Chemical Name or Material Cysteamine hydrochloride
Physical Form Solid
CAS 156-57-0
Color White
Quantity 5 g
Purity Grade Notes Research
Molecular Formula C2H8ClNS
For Use With (Application) Cancer-Kinase/protease
Synonym 2-Aminoethanethiol hydrochloride2-Mercaptoethylamine hydrochloride
Solubility Information DMSO : 250 mg/mL (2200.51 mM; Need ultrasonic) ∣H2O : ≥ 50 mg/mL (440.10 mM)
SMILES SCCN.Cl
Molecular Weight (g/mol) 113.61
Formula Weight 113.61
Percent Purity 95.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.