Learn More
CYTL1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 15856-1-AP-150UL
 View more versions of this product
Product Code. 16865234
  • 410.00 € / 150µL
Description
C17 is a cytokine-like protein specifically expressed in bone marrow and cord blood mononuclear cells that bear the CD34 surface marker (Liu et al., 2000 ).
Specifications
Specifications
CYTL1
Polyclonal
Unconjugated
CYTL1
C17, C4orf4, CYTL1, cytokine like 1, Cytokine like protein 1, Protein C17
Rabbit
Antigen Affinity Chromatography
RUO
231162, 498392, 54360
-20°C
Liquid
Immunohistochemistry (Paraffin)
0.18 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q9NRR1
CYTL1
CYTL1 Fusion Protein Ag3977
150 μL
Primary
Human, Mouse, Rat