missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Cyto-Plex™ Carboxylated Particles

Save time when measuring analytes with Thermo Scientific™ Cyto-Plex™ Carboxylated Particles, open platform particles for suspension array analysis.

Brand:  Thermo Scientific™ FM5CR01

Code : LE

Additional Details : Weight : 0.11200kg

 View more versions of this product

Product Code. 11828343

  • 171.00 € / 1mL
Estimated Shipment: 10-06-2024
to see stock.

Please to purchase this item. Need a web account? Register with us today!

Alert:

Contains sodium azide.


Description

Description

Save time when measuring analytes with Thermo Scientific™ Cyto-Plex™ Carboxylated Particles, open platform particles for suspension array analysis.

Suspension array analysis is a powerful technique enabling the simultaneous detection and quantitation of multiple analytes with a single sample. Using color coded microspheres with the appropriate dyes, these tests can now be carried out using commonly available flow cytometers.
  • Cyto-Plex™ microspheres consist of a highly uniform polystyrene particle with fluorescent intensities which are completely separated from each other
  • Carboxylated particles provide different sizes and levels of fluorescent intensities for analysis of multiple analytes
  • Use of a single diameter particle for all dye levels saves time by only requiring the development and optimization of one bead chemistry; multiple diameters can be combined to increase the number of analytes measured in one test
  • High-density binding sites and low non-specific binding enable coupling with a wide variety of antibodies, nucleic acids and other biomolecules
  • Microspheres have a maximum emission at 700 nm and can be excited with either 488 nm or 633 nm lasers
  • Emission can be collected in either the PE-Cy5 or APC channels
  • Since there is little or no emission in the FITC and PE channels, probes utilizing either of these dyes can be effectively used as reporters

Recommended for:

  • Open platform microspheres for suspension array analysis in flow cytometry
Specifications

Specifications

Sodium Azide Preservative, 0.05% Tween-20
Polystyrene microspheres in water
Red
Carboxylated Particle
Material Safety Data Sheet (MSDS) and Package Insert Sheet
1.06g/cm³
Product stable for a minimum of 12 months
≤2% CV
Refrigerate when not in use; do not freeze. Store upright and keep bottle tightly sealed. Mix product by gentle inversion by hand or vortex mixer.
Cyto-Plex Carboxylated Particles, 5µm
Polystyrene with Copolymer Grafted Surface
Firefli™ Fluorescent Red (635/700nm)
Level 1
1 mL
Carboxylate-Modified
5 μm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers