missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Cytodex 1 Gamma Microcarriers

Product Code. 15727207 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
30 g
300 g
3 kg
Unit Size:
300g
30g
3kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15727207 300 g 300g
15717207 30 g 30g
15737207 3 kg 3kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15727207 Supplier Cytiva Supplier No. 17548702
 Restricted Item
Estimated Shipment: 28-04-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Microcarrier for general use in sterilized dry powder form.

  • Cytodex 1 Gamma is designed by substituting a cross-linked dextran matrix with positively charged N, N-diethylaminoethyl (DEAE) groups distributed throughout the matrix.
  • Micro-Carriers Dry Weight: 4.3 × 106
  • Surface Area Dry Weight: 0.44 m2/g
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Applications include the production of large quantities of cells, viruses and recombinant cell products (e.g. interferon, enzymes, nucleic acids, hormones), studies on cell adhesion, diff erentiation and cell function, perfusion column culture systems, microscopy studies, harvesting mitotic cells, isolation of cells, membrane studies, storage and transport of cells, assays involving celltransfer and studies on uptage of labelled compounds
Quantity 300 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.