missing translation for 'onlineSavingsMsg'
Learn More

Cytiva Cytodex 3 Gamma Microcarriers

Product Code. 15747207 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
30 g
300 g
3 kg
Unit Size:
300g
30g
3kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15747207 30 g 30g
15767207 3 kg 3kg
15757207 300 g 300g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15747207 Supplier Cytiva Supplier No. 17548801
 Restricted Item
Estimated Shipment: 12-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

The microcarrier of choice for cells that are difficult to culture in vitro - particularly for cells with an epithelial-like morphology.

  • Cytodex 3 Gamma is designed by chemically coupling of a thin layer of denatured porcine collagen type 1 to the cross-linked dextran matrix.
  • As the collagen surface layer can be digested by a variety of proteolytic enzymes, harvest of such cells from the microcarriers can be performed with maintained high cell viability and membrane integrity.
  • This feature of Cytodex 3 Gamma is beneficial in the development of successful serial subcultivation protocols for scaling up culture volumes.
  • Applications include the production of large quantities of cells, viruses and recombinant cell products (e.g. interferon, enzymes, nucleic acids, hormones), studies on cell adhesion, differentiation and cell function, perfusion column culture systems, microscopy studies, harvesting mitotic cells, isolation of cells, membrane studies, storage and transport of cells, assays involving cell transfer and studies on uptage of labelled compounds
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Production of large quantities of cells, viruses and recombinant cell products, studies on cell adhesion, differentiation and cell function
Quantity 30 g
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.