missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytohesin 3 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
280.00 € - 572.00 €
Specifications
| Antigen | Cytohesin 3 |
|---|---|
| Dilution | Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 |
| Applications | Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18471292
|
Novus Biologicals
NBP1-90097-25ul |
25 μL |
280.00 €
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18284627
|
Novus Biologicals
NBP1-90097 |
0.1 mL |
572.00 €
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cytohesin 3 Polyclonal specifically detects Cytohesin 3 in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| Cytohesin 3 | |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Mouse | |
| ARF nucleotide-binding site opener 3, ARNO3PH, SEC7 and coiled-coil domain-containing protein 3, cytohesin 3, General receptor of phosphoinositides 1, Grp1, GRP1cytohesin-3, pleckstrin homology, Sec7 and coiled-coil domains 3, Protein ARNO3, PSCD3pleckstrin homology, Sec7 and coiled/coil domains 3 | |
| CYTH3 | |
| IgG | |
| Affinity Purified | |
| Specificity of human Cytohesin 3 antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot 0.4 ug/ml, Immunohistochemistry, Immunohistochemistry-Paraffin 1:200 - 1:500 | |
| Polyclonal | |
| Rabbit | |
| Signal Transduction | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 9265 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:DEDGGGEGGGVPEDLSLEEREELLDIRRRKKELIDDIERLKYEIAEVMTEIDNLTSVEESKTTQRNKQIAM | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title