missing translation for 'onlineSavingsMsg'
Learn More

Cytokeratin 15 Mouse, Clone: LHK15, enQuireBio™

Mouse Monoclonal Antibody

Brand:  EnquireBio 3866-MSM1-P1

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15930368

  • 462.00 € / 100µg
Estimated Shipment: 27-01-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Specifications

Specifications

Cytokeratin 15
Cytoplasmic
LHK15
Unconjugated
NP_016880103
Mouse
52kDa
Protein A/G Purified
RUO
3866
IgG2a κ
Immunohistochemistry
Monoclonal
200μg/mL
10mM PBS with 0.05% BSA and 0.05% Azide
KRT15
A 17 amino acids synthetic peptide from the C- terminal of Human Cytokeratin 15
A431 cells. Skin or Colon
100 μg
Primary
Short term: Store at 2-4°C. Long term, aliquot and store at -20°C.
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers