missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 18 Antibody (C-04 (same as Ks18.04)), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-11485PCP
This item is not returnable.
View return policy
Description
Cytokeratin 18 Monoclonal antibody specifically detects Cytokeratin 18 in Human, Mouse, Rat, Porcine, Bovine, Canine, Equine, Feline, Goat, Hamster, Ferret, Monkey, Sheep, Chicken (Negative), Xenopus (Negative) samples. It is validated for Flow Cytometry
Specifications
| Cytokeratin 18 | |
| Monoclonal | |
| PerCP | |
| PBS | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat, Pig, Bovine, Canine, Equine, Feline, Goat, Hamster, Ferret, Monkey, Sheep, Chicken (Negative), Xenopus (Negative) | |
| Purified |
| Flow Cytometry | |
| C-04 (same as Ks18.04) | |
| Flow Cytometry | |
| Cell proliferation-inducing gene 46 protein, cell proliferation-inducing protein 46, CK-18, CYK18, cytokeratin 18, cytokeratin-18, K18, keratin 18, keratin, type I cytoskeletal 18, keratin-18 | |
| Cytoskeletal preparation extracted from the human vulvar squamous carcinoma cell line A431 | |
| 0.1 mL | |
| Cancer, Cell Biology, Cellular Markers, Phospho Specific | |
| 3875 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction