missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytokeratin 19 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody has been used in 1 publication
Brand: Novus Biologicals NBP1-78278SS
This item is not returnable.
View return policy
Description
Cytokeratin 19 Polyclonal specifically detects Cytokeratin 19 in Human, Rat samples. It is validated for Western Blot, Simple Western, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| Cytokeratin 19 | |
| Polyclonal | |
| Unconjugated | |
| PBS, 30% Glycerol with 0.05% Sodium Azide | |
| CK19, CK-19, cytokeratin 19, Cytokeratin-19,40-kDa keratin intermediate filament, K19cytokeratin-19, K1CS, keratin 19, keratin, type I cytoskeletal 19, keratin, type I, 40-kd, keratin-19, MGC15366 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| IgG |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1.16 mg/mL | |
| Western Blot 2 ug/ml, Simple Western 1:3000, Immunohistochemistry 1:400, Immunocytochemistry/Immunofluorescence 1:75, Immunohistochemistry-Paraffin 1:400 | |
| P08727 | |
| KRT19 | |
| A synthetic peptide made to an N-terminal portion of the human Cytokeratin 19 protein (between residues 1-50) [UniProt P08727] | |
| 0.025 mL | |
| Cancer, Neuroscience | |
| 3880 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction