Learn More
Cytokeratin 7 Antibody (CL8723) [DyLight 488], Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-88918G
Product Code. 30308616
  • 737.00 € / 0.10mL
Description
Cytokeratin 7 Monoclonal antibody specifically detects Cytokeratin 7 in Human samples. It is validated for Immunohistochemistry,Western Blot,Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cytokeratin 7
Monoclonal
DyLight 488
CK7, CK-7, cytokeratin 7, Cytokeratin-7, K2C7, K7keratin, 55K type II cytoskeletal, keratin 7, keratin, type II cytoskeletal 7, keratin-7, MGC129731, MGC3625, Sarcolectin, SCLkeratin, simple epithelial type I, K7, type II mesothelial keratin K7, Type-II keratin Kb7
This antibody was developed using a recombinant protein derived from P08729, with the exact immunogen sequence remaining proprietary.
0.1 mL
Cell Biology, Cellular Markers
3855
Store at 4⪚C in the dark.
IgG1
Immunohistochemistry, Western Blot, Immunohistochemistry (Paraffin)
CL8723
50mM Sodium Borate
Mouse
Protein A purified
RUO
Primary
Human
Purified