Learn More
Cytokeratin, pan Antibody (K20) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-50220
 View more versions of this product
Product Code. 18695457
  • 351.00 € / 0.10mL
Description
Cytokeratin, pan Monoclonal antibody specifically detects Cytokeratin, pan in Human samples. It is validated for Western Blot, Immunohistochemistry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cytokeratin, pan
Monoclonal
1 mg/mL
Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500
AEI2;CK1;EHK;EHK1;EPPK;K1;Keratin, type II cytoskeletal 1;KRT1A,NEPPK
This Cytokeratin, pan Antibody (K20) was developed against hair keratin.
0.1 mL
Apoptosis, Cancer, Cell Biology, Cellular Markers, Signal Transduction
3848
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
IgG2a κ
Western Blot, Immunohistochemistry
K20
Unconjugated
PBS
Mouse
Protein A purified
RUO
Primary
Human
Purified