missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cytomegalovirus gB Antibody (11B10D3E4), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-06074-100ul
This item is not returnable.
View return policy
Description
Cytomegalovirus gB Monoclonal antibody specifically detects Cytomegalovirus gB in Cytomegalovirus samples. It is validated for Western Blot, ELISA
Specifications
| Cytomegalovirus gB | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
| Western Blot, ELISA | |
| 11B10D3E4 | |
| Western Blot 1:1000-1:5000, ELISA 1:1000-1:2000 | |
| CMV, CMV glycoprotein B, CMV glycoprotein GP55, Cytomegalovirus glycoprotein GP55, gB, Glycoprotein B, Glycoprotein GP55, HCMV, HCMV glycoprotein B, HCMV glycoprotein GP55, HHV 5 glycoprotein B, HHV 5 glycoprotein GP55, HHV5 glycoprotein B, HHV5 glycoprotein GP55, Human herpesvirus 5 glycoprotein B, Human herpesvirus 5 glycoprotein GP55, UL55 | |
| This antibody was produced from a hybridoma resulting from the fusion of a mouse myeloma with B cells obtained from a mouse immunized with purified, recombinant Human CMV gB (Accession#: AAA45920.1) extracellular domain (Met 1-Lys 700) linked with the cytoplasmic domain (Arg 777-Val 907). | |
| 100 μg | |
| Immunology, Infections (Virus Bacteria and Parasites), Virology Bacteria and Parasites | |
| Cytomegalovirus (CMV) | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction