Learn More
Cytonectin Polyclonal Antibody, Invitrogen™
Chicken Polyclonal Antibody
Brand: Invitrogen PA172059
Code : LD
Additional Details : Weight : 0.01000kg
Description
PA1-72059 detects Cytonectin in human samples. PA1-72059 has been successfully used in Western blot procedures. Antibody is diluted to a 1 mg/mLstock solution. This is diluted 1:1000 immediately prior to blot application. The PA1-72059 immunogen is from cytonectin isolated from a cell lysate of normal human third trimester placenta by ROOGE.
Cytonectin Polyclonal antibody specifically detects Cytonectin in Human samples. It is validated for Western Blot.Specifications
Cytonectin | |
Polyclonal | |
Unconjugated | |
Chicken | |
IgY fraction | |
RUO | |
Human | |
Antibody | |
IgY |
Western Blot | |
20 mg/mL | |
PBS with 0.05% sodium azide; pH 7.2 | |
Cytonectin isolated from a cell lysate of normal human third trimester placenta by ROOGE. | |
10 mg | |
Primary | |
-20° C, Avoid Freeze/Thaw Cycles | |
Liquid |