missing translation for 'onlineSavingsMsg'
Learn More

D-Alpha-Methyl DOPA, TRC

Product Code. 30480794
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30480794 25 mg 25mg
30476280 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30480794 Supplier Toronto Research Chemicals Supplier No. TRCM30379025MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2799-15-7
Molecular Formula C10H13NO4
Molecular Weight (g/mol) 211.21
Synonym D-Tyrosine, 3-hydroxy-α-methyl-, Alanine, 3-(3,4-dihydroxyphenyl)-2-methyl-, D- (8CI), 3-Hydroxy-α-methyl-D-tyrosine, (+)-α-Methyldopa, D-(3,4-Dihydroxyphenyl)-2-methylalanine, D-Methyldopa, D-α-Methyl-3-(3,4-dihydroxyphenyl)alanine, D-α-Methyldopa
IUPAC Name (2R)-2-amino-3-(3,4-dihydroxyphenyl)-2-methylpropanoic acid
SMILES C[C@@](N)(Cc1ccc(O)c(O)c1)C(=O)O

Specifications

Quantity 25 mg
InChI Formula InChI=1S/C10H13NO4/c1-10(11,9(14)15)5-6-2-3-7(12)8(13)4-6/h2-4,12-13H,5,11H2,1H3,(H,14,15)/t10-/m1/s1
Formula Weight 211.0845
Chemical Name or Material D-a-Methyl DOPA
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.