missing translation for 'onlineSavingsMsg'
Learn More

D-(+)-Epinephrine, TRC

Product Code. 30012429
Change view
Click to view available options
Quantity:
10 mg
100 mg
Unit Size:
100mg
10mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012429 10 mg 10mg
30004424 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30012429 Supplier Toronto Research Chemicals Supplier No. TRCE58857510MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 150-05-0
Molecular Formula C9 H13 N O3
Molecular Weight (g/mol) 183.2
Synonym 1,2-Benzenediol, 4-[(1S)-1-hydroxy-2-(methylamino)ethyl]-, 1,2-Benzenediol, 4-[1-hydroxy-2-(methylamino)ethyl]-, (S)-, Benzyl alcohol, 3,4-dihydroxy-α-[(methylamino)methyl]-, (+)- (8CI), 4-[(1S)-1-Hydroxy-2-(methylamino)ethyl]-1,2-benzenediol, (+)-(S)-Epinephrine, (+)-1-(3,4-Dihydroxyphenyl)-2-methylaminoethanol, (+)-Adrenaline, (+)-Epinephrine, (S)-Adrenaline, (S)-Epinephrine, D(+)-Adrenaline, D-Adrenaline, D-Epinephrine, d-Adrenaline, d-Epinephrine
IUPAC Name 4-[(1S)-1-hydroxy-2-(methylamino)ethyl]benzene-1,2-diol
SMILES CNC[C@@H](O)c1ccc(O)c(O)c1

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C9H13NO3/c1-10-5-9(13)6-2-3-7(11)8(12)4-6/h2-4,9-13H,5H2,1H3/t9-/m1/s1
Formula Weight 183.0895
Percent Purity >95%
Chemical Name or Material D-(+)-Epinephrine
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.