missing translation for 'onlineSavingsMsg'
Learn More

D-Galacturonic Acid Monohydrate, TRC

Product Code. 30009696
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30009696 5 g 5g
30005601 25 g 25g
30012759 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30009696 Supplier Toronto Research Chemicals Supplier No. TRCG1865005G
 Restricted Item
Estimated Shipment: 07-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91510-62-2
Molecular Formula C6 H10 O7 . H2 O
Molecular Weight (g/mol) 212.15
Synonym D-Galacturonic Acid Hydrate (1:1), D-Galacturonic Acid, Monohydrate (9CI)
IUPAC Name (2S,3R,4S,5R)-2,3,4,5-tetrahydroxy-6-oxohexanoic acid;hydrate
SMILES O.O[C@@H](C=O)[C@@H](O)[C@@H](O)[C@H](O)C(=O)O

Specifications

Quantity 5 g
InChI Formula InChI=1S/C6H10O7.H2O/c7-1-2(8)3(9)4(10)5(11)6(12)13;/h1-5,8-11H,(H,12,13);1H2/t2-,3+,4+,5-;/m0./s1
Formula Weight 212.0532
Percent Purity >95%
Chemical Name or Material D-Galacturonic acid hydrate
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.