missing translation for 'onlineSavingsMsg'
Learn More

D-gamma-Tocopherol, 95%

Product Code. 10279990 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 mg
Packaging:
Glass bottle
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10279990 100 mg Glass bottle 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10279990 Supplier Thermo Scientific Acros Supplier No. 421041000
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 54-28-4 | C28H48O2 | 416.69 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

CAS 54-28-4
Color Brown
Boiling Point 200°C (0.1 mmHg)
Flash Point >93°C
Infrared Spectrum Authentic
Assay Percent Range 94% min. (HPLC)
Packaging Glass bottle
Molecular Formula C28H48O2
MDL Number MFCD00066529
Quantity 100 mg
Merck Index 15, 9655
Specific Rotation Condition +30° (25°C c=2.00 in isooctane)
Specific Rotation + 30
Synonym gamma-tocopherol, +-gamma-tocopherol, d-gamma-tocopherol, 7,8-dimethyltocol, unii-8ef1z1238f, rrr-gamma-tocopherol, r,r,r-gamma-tocopherol, 2r-2,7,8-trimethyl-2-4r,8r-4,8,12-trimethyltridecyl chroman-6-ol, 2r-2,7,8-trimethyl-2-4r,8r-4,8,12-trimethyltridecyl-3,4-dihydro-2h-chromen-6-ol, 2r-3,4-dihydro-2,7,8-trimethyl-2-4r,8r-4,8,12-trimethyltridecyl-2h-1-benzopyran-6-ol
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol,acetone,chloroform,ether,,oils,fats
InChI Key QUEDXNHFTDJVIY-DQCZWYHMSA-N
SMILES CC1=C(C=C2CCC(OC2=C1C)(C)CCCC(C)CCCC(C)CCCC(C)C)O
IUPAC Name (2R)-2,7,8-trimethyl-2-[(4R,8R)-4,8,12-trimethyltridecyl]-3,4-dihydrochromen-6-ol
Molecular Weight (g/mol) 416.69
PubChem CID 92729
ChEBI CHEBI:18185
Formula Weight 416.69
Percent Purity 95%
Physical Form Viscous Liquid
Chemical Name or Material D-γ-Tocopherol, 95%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.