missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-Glucose-6-phosphate disodium salt hydrate, 98%

Product Code. 15457219
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
Product Code. Quantity unitSize
15457219 1 g 1g
15467219 5 g 5g
2 options
This item is not returnable. View return policy

Product Code. 15457219

Brand: Thermo Scientific Chemicals J66023.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 3671-99-6 | C6H11Na2O9P | 304.10 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3671-99-6
Molecular Formula C6H11Na2O9P
Molecular Weight (g/mol) 304.10
MDL Number MFCD00136037
InChI Key CHIBNKGHYAQTQY-OGPXQWFKNA-L
Synonym d-glucose-6-phosphate disodium salt, sodium 2r,3r,4s,5r-2,3,4,5-tetrahydroxy-6-oxohexyl phosphate, sodium glucose 6-phosphate, disodium d-glucose 6-phosphate, glucose 6-phosphate sodium salt, glucose-6-phosphate disodium, disodium d-glucose-6-phosphate, d-glucose 6-disodium phosphate, d-glucose 6-phosphoric acid disodium salt, disodium 2r,3s,4r,5r-2,3,4,5-tetrahydroxy-6-phosphonooxy hexanal
PubChem CID 18594434
IUPAC Name disodium;[(2R,3R,4S,5R)-2,3,4,5-tetrahydroxy-6-oxohexyl] phosphate
SMILES [Na+].[Na+].O[C@H]1O[C@H](COP([O-])([O-])=O)[C@@H](O)[C@H](O)[C@H]1O

Specifications

Color White
Quantity 1 g
Solubility Information Soluble in water
Formula Weight 304.1
Percent Purity 98%
Physical Form Powder
Chemical Name or Material D-Glucose-6-phosphate disodium salt

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.