missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-(+)-Glucose monohydrate

Product Code. 11408963
Change view
Click to view available options
Quantity:
500 g
2500 g
Unit Size:
2500g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11408963 2500 g 2500g
11498953 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11408963 Supplier Thermo Scientific Chemicals Supplier No. A11090.0E
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 14431-43-7 | C6H12O6·H2O

D-(+)-Glucose monohydrate is used by industry professionals to indicate the degree to which a substance contains water, and in food industry, pharmaceuticals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-(+)-Glucose monohydrate is used by industry professionals to indicate the degree to which a substance contains water, and in food industry, pharmaceuticals.

Solubility
Soluble in water.

Notes
Hygroscopic. Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14431-43-7
Molecular Formula C6H12O6·H2O
MDL Number MFCD00149450

Specifications

Melting Point ∼83°C
Assay Percent Range 99%
Quantity 2500 g
Solubility Information Soluble in water.
Formula Weight 198.18 (180.16 Anhydrous)
Chemical Name or Material D- (+)-Glucose monohydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.