accessibility menu, dialog, popup

UserName

H2OHO(R)O(S)OH(R)OH(S)HO(S)HO

CAS RN 14431-43-7

IUPAC Name: (2S,3R,4S,5S,6R)-6-(hydroxymethyl)oxane-2,3,4,5-tetrol hydrate
Synonyms: α-glucose hydrate
Molecular Weight (g/mol): 198.17
Molecular Formula: C6H14O7
InChi Key: OSNSWKAZFASRNG-WNFIKIDCSA-N
SMILES: O.OC[C@H]1O[C@H](O)[C@H](O)[C@@H](O)[C@@H]1O
Quantity 
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
Grade 
  • (3)
Certifications/Compliance 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (1)
  • (1)
  • (3)
  • (2)
  • (3)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)
  • (3)
  • (1)
  • (1)
  • (2)
  • (1)