missing translation for 'onlineSavingsMsg'
Learn More

D(+)-Glucose monohydrate, 99+%, extra pure

Product Code. 15294844 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 kg
5 kg
25 kg
Packaging:
Fibre drum
Plastic Bottle
Plastic Drum
Unit Size:
1kg
25kg
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
15294844 25 kg Fibre drum 25kg
15274844 1 kg Plastic Bottle 1kg
15284844 5 kg Plastic Drum 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15294844 Supplier Thermo Scientific Acros Supplier No. 450740250
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 14431-43-7 | C6H12O6·H2O | 198.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14431-43-7
Molecular Formula C6H12O6·H2O
Molecular Weight (g/mol) 198.17

Specifications

Ignition Residue 0.1% max
Assay Percent Range 99% min. (ELSD) (HPLC)
Packaging Fibre drum
Quantity 25 kg
Solubility Information Solubility in water: soluble.
Formula Weight 198.17
Percent Purity 99+%
Grade Extra Pure
Chemical Name or Material D(+)-Glucose monohydrate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.