missing translation for 'onlineSavingsMsg'
Learn More

D(+)-Glucose Monohydrate, Extra Pure, SLR, meets analytical specification of Ph.Eur., BP, Fisher Chemical™

Product Code. 11463743 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1 kg
5 kg
Packaging:
Plastic keg
Plastic powder jar
Unit Size:
1kg
5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11463743 5 kg Plastic keg 5kg
10529190 1 kg Plastic powder jar 1kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11463743 Supplier Fisher Chemical Supplier No. G/0400/65
Request Bulk or Custom Format

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

D(+)-Glucose Monohydrate, Extra Pure, SLR, meets analytical specification of Ph.Eur., BP

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14431-43-7
Molecular Formula C6H12O6·H2O
MDL Number 149450

Specifications

Color White
pH 5.9
Packaging Plastic keg
Quantity 5 kg
Formula Weight 198.18g/mol
Physical Form Solid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.