missing translation for 'onlineSavingsMsg'
Learn More

D,L-Carbidopa, TRC

Product Code. 30362646
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
50 mg
Unit Size:
2.50mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30362646 50 mg 50mg
30360945 2.5 mg 2.50mg
30360167 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30362646 Supplier Toronto Research Chemicals Supplier No. TRCC17591550MG
 Restricted Item
Estimated Shipment: 14-09-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 302-53-4
Molecular Formula C10 H14 N2 O4
Molecular Weight (g/mol) 226.23
Synonym Benzenepropanoic acid, α-hydrazinyl-3,4-dihydroxy-α-methyl-, Benzenepropanoic acid, α-hydrazino-3,4-dihydroxy-α-methyl- (9CI), Hydrocinnamic acid, α-hydrazino-3,4-dihydroxy-α-methyl- (7CI,8CI), α-Hydrazinyl-3,4-dihydroxy-α-methylbenzenepropanoic acid, DL-3-(3,4-Dihydroxyphenyl)-2-hydrazino-2-methylpropionic acid, DL-Carbidopa, DL-α-Hydrazino-3,4-dihydroxy-α-methylhydrocinnamic acid, DL-α-Hydrazino-α-methyl-3,4-dihydroxyphenylpropionic acid, DL-α-Hydrazino-α-methyl-β-(3,4-dihydroxyphenyl)propionic acid, DL-α-Hydrazino-α-methyldopa, DL-α-MDH, NSC 92521, dl-MK 485, dl-α-Methyl-α-hydrazino-3,4-dihydroxyphenylpropionic acid, α-Hydrazino-α-methyl-3,4-dihydroxyphenylpropionic acid, α-Hydrazino-α-methyldopa, α-MDH, α-Methylhydrazinodopa, β-(3,4-Dihydroxyphenyl)-α-hydrazino-α-methylpropionic acid, β-(3,4-Dihydroxyphenyl)-α-methyl-α-hydrazinopropionic acid
IUPAC Name 3-(3,4-dihydroxyphenyl)-2-hydrazinyl-2-methylpropanoic acid
SMILES CC(Cc1ccc(O)c(O)c1)(NN)C(=O)O

Specifications

Quantity 50 mg
InChI Formula InChI=1S/C10H14N2O4/c1-10(12-11,9(15)16)5-6-2-3-7(13)8(14)4-6/h2-4,12-14H,5,11H2,1H3,(H,15,16)
Formula Weight 226.0954
Chemical Name or Material D,L Carbidopa
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.