missing translation for 'onlineSavingsMsg'
Learn More

D-Lactose 1000 μg/mL in Water, Dr. Ehrenstorfer

Product Code. 30315788
Change view
Click to view available options
Quantity:
1 mL
Unit Size:
1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30315788 1 mL 1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30315788 Supplier Dr. Ehrenstorfer Supplier No. DREA14590800WA1000
 Restricted Item
Estimated Shipment: 17-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Discover Dr. Ehrenstorfer’s certified reference materials: available in multiple formats, including multi-component regulatory mixtures, to power your food and environmental analysis with traceable, ISO-accredited quality

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material Lactose
CAS 63-42-3
Analyte or Component Names Lactose
Shipping Condition Room Temperature
Molecular Formula C12 H22 O11
Concentration 1000 μg/mL
Certifications/Compliance ISO 17034
Synonym Lactose, O-β-d-Galactopyranosyl-(1→4)-β-d-glucopyranose, D-Glucose, 4-O-β-D-galactopyranosyl-, Lactose (8CI), (+)-Lactose, AHL, Aletobiose, D-(+)-Lactose, DCL 15, DCl 11, Dairylac 80, Dilactose, Fast-Flo Lactose, Fast-flo, Flowlac 100, Galactinum, Granulac 140, Granulac 140M, Granulac 70, Inhalac 250, Inhalac 70, LE-PRO LAC 451, Lactin, Lactin (carbohydrate), Lactobiose, Lactohale 100, Lactohale 200, Lactohale 300, Lactose DCL 11, Lactose Edible 90, Lactose Fast-flo, Lactose anhydride, Lactose anhydrous, Milk sugar, Nonpareil 107, Osmolactan, Pharmatosa DCL 21, Pharmatose 21, Pharmatose 325M, Pharmatose DCL 15, PrenoLakt, Prismalac, Respitose MC 001, Respitose ML 001, Respitose ML 003, Respitose ML 006, Respitose SV 003, Respitose SV 010, Saccharum lactin, Sachelac, Sorbalac 400, Sorbolac 400, Spherolac, Super-Tab, SuperTab 11SD, SuperTab 21AN, Tablettose, Tablettose 70, Tablettose 80, Variolac 960, Zeparox EP
SMILES OC[C@H]1O[C@@H](O[C@H]2[C@H](O)[C@@H](O)[C@H](O)O[C@@H]2CO)[C@H](O)[C@@H](O)[C@H]1O
IUPAC Name (2R,3R,4S,5R,6S)-2-(hydroxymethyl)-6-[(2R,3S,4R,5R,6RS)-4,5,6-trihydroxy-2-(hydroxymethyl)tetrahydropyran-3-yl]oxy-tetrahydropyran-3,4,5-triol
Solvent or Matrix Water
Molecular Weight (g/mol) 342.3
Quantity 1 mL
Formula Weight 342.1162 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.