missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-Methionine, 99%

Product Code. 11484717
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11484717 100 g 100g
11464717 5 g 5g
11474717 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 11484717 Supplier Thermo Scientific Chemicals Supplier No. B21213.22
 Restricted Item
In Stock
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 348-67-4 | C5H11NO2S | 149.21 g/mol

D-Methionine is used to rescue noise-induced hearing loss. It is used to improve the nutritive value of animal feed.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
D-Methionine is used to rescue noise-induced hearing loss. It is used to improve the nutritive value of animal feed.

Solubility
Soluble in water. 53 g/L (20°C).

Notes
Store away from oxidizing agents. Store in a cool, dry condition in a well sealed container.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 348-67-4
Molecular Formula C5H11NO2S
Molecular Weight (g/mol) 149.21
MDL Number MFCD00002622
InChI Key FFEARJCKVFRZRR-SCSAIBSYSA-N
Synonym d-methionine, h-d-met-oh, r-methionine, d-methionin, d-metionien, methionine, d, r-2-amino-4-methylthio butanoic acid, 2r-2-amino-4-methylsulfanyl butanoic acid, d-2-amino-4-methylthio butyric acid
PubChem CID 84815
ChEBI CHEBI:16867
IUPAC Name (2R)-2-amino-4-methylsulfanylbutanoic acid
SMILES CSCC[C@@H](N)C(O)=O

Specifications

Melting Point ∼280°C (decomposition)
Beilstein 1722293
Merck Index 14,5975
Solubility Information Soluble in water. 53g/L (20°C).
Quantity 100 g
Formula Weight 149.21
Optical Rotation −23° (c=2 in 2N HCl)
Percent Purity 99%
Chemical Name or Material D-Methionine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.