accessibility menu, dialog, popup

UserName

CAS RN 348-67-4

IUPAC Name: (2R)-2-amino-4-(methylsulfanyl)butanoic acid
Synonyms: D-methionine poly-l-methionine meonine liquimeth DL-isomer methionine methionine, DL isomer urimeth D-methionin DL isomer methionine racemethionine
Molecular Weight (g/mol): 149.21
Molecular Formula: C5H11NO2S
InChi Key: FFEARJCKVFRZRR-SCSAIBSYSA-N
SMILES: CSCC[C@@H](N)C(O)=O
Quantity 
  • (1)
  • (3)
  • (3)
  • (1)
  • (2)
Percent Purity 
  • (3)
  • (1)
  • (3)
Color 
  • (1)
Grade 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results