missing translation for 'onlineSavingsMsg'
Learn More

(D)-(+)-Neopterin, Tocris Bioscience™

Product Code. 30001681
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 30001681

Brand: Tocris Bioscience 4656/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Stimulated by IFN-γ; marker of immune activation

Precursor of biopterin synthesis. Synthesized in response to interferon-γ stimulation; used as a marker of T helper cell-induced immune activation. Also an indicator of oxidative stress; modulates the effects of reactive oxygen species (ROS).
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2009-64-5
Molecular Formula C9H11N5O4
Molecular Weight (g/mol) 253.218
InChI Key BMQYVXCPAOLZOK-XINAWCOVSA-N
Synonym d-+-neopterin, neopterin, d-neopterin, d-erythro-neopterin, 6-d-erythro-neopterin, l-neopterin, unii-9pld60n7so, 9pld60n7so, l-erythro-neopterin, 1s,2r-1-2-amino-4-hydroxypteridin-6-yl propane-1,2,3-triol
PubChem CID 448839
IUPAC Name 2-amino-6-[(1S,2R)-1,2,3-trihydroxypropyl]-1H-pteridin-4-one
SMILES C1=C(N=C2C(=N1)NC(=NC2=O)N)C(C(CO)O)O

Specifications

Quantity 10 mg
Formula Weight 253.21
Percent Purity >98%
Chemical Name or Material (D)-(+)-Neopterin
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.