missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D-(-)-Norleucine, 99%

Product Code. 11348065
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
This item is not returnable. View return policy

Product Code. 11348065

Brand: Thermo Scientific Chemicals L08257.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 327-56-0 | C6H13NO2 | 131.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 327-56-0
Molecular Formula C6H13NO2
Molecular Weight (g/mol) 131.18
MDL Number MFCD00008099
InChI Key LRQKBLKVPFOOQJ-RXMQYKEDSA-N
Synonym d-norleucine, h-d-nle-oh, r-2-aminohexanoic acid, d-2-aminohexanoic acid, d---norleucine, 2r-2-aminohexanoic acid, unii-vp8g7lx265, norleucine, norleucine, d--, d-nor-leucine
PubChem CID 456468
ChEBI CHEBI:42101
IUPAC Name (2R)-2-aminohexanoic acid
SMILES CCCC[C@@H](N)C(O)=O

Specifications

Melting Point >300°C
Beilstein 1721749
Merck Index 14,6706
Quantity 1 g
Formula Weight 131.18
Optical Rotation −23° (c=5 in 5N HCl)
Percent Purity 99%
Chemical Name or Material D-(-)-Norleucine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.