missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D-Pantothenic acid sodium, MedChemExpress
Description
D-Pantothenic acid sodium (Sodium pantothenate) is an essential trace nutrient that functions as the obligate precursor of coenzyme A (CoA). D-Pantothenic acid sodium plays key roles in myriad biological processes, including many that regulate carbohydrate, lipid, protein, and nucleic acid metabolism.
Specifications
Specifications
| Chemical Name or Material | D-Pantothenic acid sodium |
| Physical Form | Solid |
| CAS | 867-81-2 |
| Color | White |
| Quantity | 100 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C9H16NNaO5 |
| Synonym | Sodium pantothenateVitamin B5 sodium |
| Solubility Information | H2O : 100 mg/mL (414.56 mM; Need ultrasonic) ∣DMSO : 5 mg/mL (20.73 mM; Need ultrasonic) |
| SMILES | OCC(C)(C)[C@@H](O)C(NCCC(O[Na])=O)=O |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?