missing translation for 'onlineSavingsMsg'
Learn More

D-Proline, TRC

Product Code. 30423917
Change view
Click to view available options
Quantity:
100 mg
1 g
5 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30423917 1 g
30425926 5 g
30427186 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30423917 Supplier Toronto Research Chemicals Supplier No. TRCP7559901G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 344-25-2
Molecular Formula C5 H9 N O2
Molecular Weight (g/mol) 115.13
Synonym D-Proline, Proline, D- (8CI), (+)-(R)-Proline, (R)-(+)-Proline, (R)-2-Carboxypyrrolidine, (R)-Proline, (R)-Pyrrolidine-2-carboxylic acid, (R)-Pyrrolidine-2-carboxylic acid, D-(+)-Proline
IUPAC Name (2R)-pyrrolidine-2-carboxylic acid
SMILES OC(=O)[C@H]1CCCN1

Specifications

InChI Formula InChI=1S/C5H9NO2/c7-5(8)4-2-1-3-6-4/h4,6H,1-3H2,(H,7,8)/t4-/m1/s1
Quantity 1 g
Formula Weight 115.0633
Shipping Condition Room Temperature
Chemical Name or Material D-Proline
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.