missing translation for 'onlineSavingsMsg'
Learn More

D-Sorbitol, MedChemExpress

Product Code. 16448286
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16448286 100 mg 100mg
16402126 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16448286 Supplier Medchem Express Supplier No. HYB0400
 Restricted Item
Estimated Shipment: 08-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress D-Sorbitol (Sorbitol) is a six-carbon sugar alcohol and can used as a sugar substitute. D-Sorbitol can be used as a stabilizing excipient and/or isotonicity agent, sweetener, humectant, thickener and dietary supplement.

D-Sorbitol (Sorbitol) is a six-carbon sugar alcohol and can used as a sugar substitute. D-Sorbitol can be used as a stabilizing excipient and/or isotonicity agent, sweetener, humectant, thickener and dietary supplement.

Specifications

Chemical Name or Material D-Sorbitol
Physical Form Solid
CAS 50-70-4
Color White
Quantity 100 mg
Purity Grade Notes Research
Molecular Formula C6H14O6
Synonym Sorbitol D-Glucitol
Solubility Information H2O : 100 mg/mL (548.94 mM; Need ultrasonic) ∣DMSO : 100 mg/mL (548.94 mM; Need ultrasonic)
SMILES OC[C@@H]([C@H]([C@@H]([C@@H](CO)O)O)O)O
Molecular Weight (g/mol) 182.17
Formula Weight 182.17
Percent Purity 98.0%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.