missing translation for 'onlineSavingsMsg'
Learn More

D-tert-Leucine Methyl Ester Hydrochloride, TRC

Product Code. 30349641
Change view
Click to view available options
Quantity:
500 mg
1 g
2 g
Unit Size:
1g
2g
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30349641 500 mg 500mg
30353624 1 g 1g
30360040 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30349641 Supplier Toronto Research Chemicals Supplier No. TRCD246930500MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 167223-43-0
Molecular Formula C7H15NO2 . HCl
Molecular Weight (g/mol) 181.66
IUPAC Name methyl (2R)-2-amino-3,3-dimethylbutanoate;hydrochloride
SMILES Cl.COC(=O)[C@H](N)C(C)(C)C

Specifications

InChI Formula InChI=1S/C7H15NO2.ClH/c1-7(2,3)5(8)6(9)10-4;/h5H,8H2,1-4H3;1H/t5-;/m0./s1
Quantity 500 mg
Formula Weight 181.087
Shipping Condition Room Temperature
Chemical Name or Material D-tert-Leucine Methyl Ester Hydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.