missing translation for 'onlineSavingsMsg'
Learn More

D-delta-Tocopherol, 93%

Product Code. 10391641 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mg
500 mg
Packaging:
Glass bottle
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 10391641

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 119-13-1 | C27H46O2 | 402.66 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 119-13-1
Molecular Formula C27H46O2
Molecular Weight (g/mol) 402.66
MDL Number MFCD20486794
InChI Key GZIFEOYASATJEH-VHFRWLAGSA-N
Synonym delta-tocopherol, unii-r0zb2556p8 component, unii-zl0rfa4e5n component, s-2,8-dimethyl-2-4s,8s-4,8,12-trimethyltridecyl-3,4-dihydro-2h-1-benzopyran-6-ol, 2s-2,8-dimethyl-2-4s,8s-4,8,12-trimethyltridecyl-3,4-dihydro-1-benzopyran-6-ol, 2h-1-benzopyran-6-ol,3,4-dihydro-2,8-dimethyl-2-4r,8r-4,8,12-trimethyltridecyl-, 2r
PubChem CID 12444418
SMILES CC(C)CCC[C@@H](C)CCC[C@@H](C)CCC[C@]1(C)CCC2=CC(O)=CC(C)=C2O1

Specifications

Color Brown to Yellow
Flash Point >93°C
Infrared Spectrum Authentic
Assay Percent Range 91% min. (GC)
Packaging Glass bottle
Quantity 100 mg
Merck Index 15,9656
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in ethanol, acetone, chloroform, ether,, oils, fats
IUPAC Name (2S)-2,8-dimethyl-2-[(4S,8S)-4,8,12-trimethyltridecyl]-3,4-dihydrochromen-6-ol
Formula Weight 402.65
Percent Purity 93%
Physical Form Viscous Liquid
Chemical Name or Material D-δ-Tocopherol
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.