missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals D(+)-Xylose, 99+%

Product Code. 10113100 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Unit Size:
100g
2.5kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10113100 100 g Plastic bottle 100g
10487720 500 g Plastic bottle 500g
11916901 2.5 kg Plastic bottle 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10113100 Supplier Thermo Scientific Chemicals Supplier No. 141001000
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 58-86-6 | C5H10O5 | 150.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 58-86-6
Molecular Formula C5H10O5
Molecular Weight (g/mol) 150.13

Specifications

Melting Point 148°C to 153°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 99% min. (ELSD) (HPLC)
Packaging Plastic bottle
Quantity 100 g
Beilstein 31,47
Merck Index 15,10285
Specific Rotation Condition + 20.00 (20.00°C c=10,H2O)
Specific Rotation + 20
Solubility Information Solubility in water: soluble. Other solubilities: soluble in pyridine and hot alcohol
Formula Weight 150.13
Percent Purity 99+%
Physical Form Fine Crystalline Powder
Chemical Name or Material D(+)-Xylose
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.