accessibility menu, dialog, popup

UserName

CAS RN 58-86-6

IUPAC Name: oxane-2,3,4,5-tetrol
Synonyms: xylose
Molecular Weight (g/mol): 150.13
Molecular Formula: C5H10O5
InChi Key: SRBFZHDQGSBBOR-UHFFFAOYNA-N
SMILES: OC1COC(O)C(O)C1O
Quantity 
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
Percent Purity 
  • (4)
  • (3)
Physical Form 
  • (2)
  • (3)
  • (1)
  • (1)
Grade 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (3)
  • (6)
  • (3)
  • (1)
  • (3)
  • (1)
  • (1)
  • (1)
  • (3)
  • (4)
  • (3)
  • (2)
  • (3)
  • (1)
  • (1)
  • (2)