missing translation for 'onlineSavingsMsg'
Learn More

D(+)-Xylose, C.P. grade, 99% max., MP Biomedicals™

Product Code. 11417531 Shop All MP Biomedicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11417531 500 g 500g
11407531 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11417531 Supplier MP Biomedicals Supplier No. 0210330090
  • ONLINE EXCLUSIVE PRICE
    256.00 €
    242.55 € / 500g
    Save 13.45 €
    5% Off
 Restricted Item
Estimated Shipment: 17-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Chemical Identifiers

CAS 58-86-6
Molecular Formula C5H10O5
MDL Number MFCD00064360
Synonym Wood sugar

Specifications

Color White
Assay Percent Range >99%
Quantity 500 g
Formula Weight 150.13
Grade C.P.
Physical Form Crystalline Powder
Chemical Name or Material D(+)-Xylose
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.