missing translation for 'onlineSavingsMsg'
Learn More
Learn More
D2-40 Mouse Monoclonal Antibody, Epredia™
Obtain accurate, reproducible results in immunohistochemistry experiments with Epredia™ D2-40 Mouse Monoclonal Antibody.
Brand: Epredia MS-1940-P0
This item is not returnable.
View return policy
Description
D2-40 is reactive to a 40kDa O-linked sialoglycoprotein in lymphatic endothelium, fetal testis, seminomas and dysgerminomas. D2-40 is useful in demonstrating lymphatic invasion by primary tumors and in vascular lesions. Publications suggest that D2-40 positivity in lymphovascular invasion was a more prognostic predictor of breast cancer than blood vessel invasion2. It is also considered useful in differential diagnosis of epithelioid malignant mesothelioma vs adenocarcinoma1.Host Species: Mouse
Clone: D2-40
Isotype: IgG1
Species Reactivity: Human. Others not tested
Epitope: Not determined
Immunogen: O-linked sialoglycoprotein
Molecular Weight: 40kDa
Positive Control: Breast carcinomas, mesothelioma and colon carcinoma
Cellular Localization: Membranous/Cytoplasmic
Recommended for: Immunohistochemistry
Specifications
| Immunohistochemistry | |
| Monoclonal | |
| Unconjugated | |
| Mouse | |
| 40kDa | |
| 0.1 mL | |
| Primary | |
| Store vial at 4°C. When stored at 2-8°C, this antibody is stable for 24 months. | |
| IgG1 |
| Membranous/Cytoplasmic | |
| D2-401 | |
| Antibody purified from tissue culture supernatant. Prepared in 10mM PBS, pH 7.4, with 0.2% BSA and <0.1% sodium azide. | |
| O-linked sialoglycoprotein | |
| Breast carcinomas, mesothelioma and colon carcinoma | |
| RUO | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction